N-Acetyl-D-phenylalanin

APOSOR322104-5GEA 0 EUR
APOSOR322104-5G
N-Acetyl-D-phenylalanin
N-Acetyl-L-phenylalanine
Formel: C₁₁H₁₃NO₃
Molekulargewicht: 207,2 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00063158
CAS-Nummer: 2018-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR