2-Amino-3-formylchromon
APOSOR322190-1GEA
0
EUR
APOSOR322190-1G
2-Amino-3-formylchromon
2-Amino-3-formylchromon
Formel:
C₁₀H₇NO₃ Molekulargewicht: 189,1675 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191735 CAS-Nummer: 61424-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...