4'-(4-Methyl-1-piperazinyl)acetophenon
APOSOR322361-500MGEA
0
EUR
APOSOR322361-500MG
4'-(4-Methyl-1-piperazinyl)acetophenon
4'-(4-Methyl-1-piperazinyl)acetophenon
Formel:
C₁₃H₁₈N₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00023297 CAS-Nummer: 26586-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...