2-Chlor-4-iodnicotinsäure
APOSOR322373-500MGEA
0
EUR
APOSOR322373-500MG
2-Chlor-4-iodnicotinsäure
2-Chlor-4-iodnicotinsäure
Formel:
C₆H₃ClINO₂ Molekulargewicht: 283,45 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05663520 CAS-Nummer: 544671-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...