(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%
APOSOR322413-5GEA
0
EUR
APOSOR322413-5G
APOSOR322413-25G
APOSOR322413-1G
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol
Formel:
C₁₁H₁₅NO₂ Molekulargewicht: 193,2423 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01073893 CAS-Nummer: 90365-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...