(S)-(+)-2-Nonylisothiocyanat
APOSOR322589-1GEA
0
EUR
APOSOR322589-1G
(S)-(+)-2-Nonylisothiocyanat
(S)-(+)-2-Nonylisothiocyanat
Formel:
C₁₀H₁₉NS Molekulargewicht: 185,33 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04039335 CAS-Nummer: 737000-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...