3-(3-Bromphenylsulfonamido)phenylboronsäure 95%
APOSOR3335-5GEA
0
EUR
APOSOR3335-5G
APOSOR3335-1G
3-(3-Bromphenylsulfonamido)phenylboronsäure 95%
3-(3-Bromphenylsulfonamido)phenylboronsäure
Formel:
C₁₂H₁₁BBrNO₄S Molekulargewicht: 356,0000 g/mol Schmelzpunkt: 97…100 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08436047 CAS-Nummer: 913835-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...