4-Methoxy-2-nitrobenzonitril 95%
Lieferant: Apollo Scientific
APOSOR345089-25GEA
207
EUR
APOSOR345089-25G
4-Methoxy-2-nitrobenzonitril 95%
4-Methoxy-2-nitrobenzonitril
Formel:
C₈H₆N₂O₃ Molekulargewicht: 178,1500 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00667741 CAS-Nummer: 38469-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...