5-Chlor-3-methylanthranilsäure
Lieferant: Apollo Scientific
APOSOR350000-100GEA
80
EUR
APOSOR350000-100G
5-Chlor-3-methylanthranilsäure
5-Chlor-3-methylanthranilsäure
Formel:
C₈H₈ClNO₂ Molekulargewicht: 185,6076 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02358895 CAS-Nummer: 20776-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...