3,5-Dibromchinolin

APOSOR350124-1GEA 0 EUR
APOSOR350124-1G
3,5-Dibromchinolin
3,5-Dibromoquinoline
Formel: C₉H₅Br₂N
Molekulargewicht: 286,95 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12024477
CAS-Nummer: 101861-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR