2,3-Dimethyl-6-nitro-1H-indazol

Lieferant: Apollo Scientific

APOSOR350127-1GEA 902 EUR
APOSOR350127-1G
2,3-Dimethyl-6-nitro-1H-indazol
2,3-Dimethyl-6-nitro-1H-indazole
Formel: C₉H₁₁N₃O₂
Molekulargewicht: 191,1867 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10703326
CAS-Nummer: 444731-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR