2-Ethoxybenzonitril

Lieferant: Apollo Scientific

APOSOR350161-25GEA 181 EUR
APOSOR350161-25G
2-Ethoxybenzonitril
2-Ethoxybenzonitril
Formel: C₉H₉NO
Molekulargewicht: 147,18
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00001789
CAS-Nummer: 6609-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR