(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-4-[(2-methyl-2-propanyl)oxy]-4-oxobutansäure
APOSOR350180-25GEA
0
EUR
APOSOR350180-25G
(2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-4-[(2-methyl-2-propanyl)oxy]-4-oxobutansäure
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
Formel:
C₂₃H₂₅NO₆ Molekulargewicht: 411,5 g/mol Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
71989-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...