Methyl-2-(4-(tert-butyl)phenyl)acetate

Lieferant: Apollo Scientific

APOSOR350246-100GEA 112 EUR
APOSOR350246-100G
Methyl-2-(4-(tert-butyl)phenyl)acetate
Methyl-2-(4-(tert-butyl)phenyl)acetate
Formel: C₁₃H₁₈O₂
Molekulargewicht: 206,29 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00051913
CAS-Nummer: 3549-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR