3-(Methoxycarbonyl)benzoesäure
Lieferant: Apollo Scientific
APOSOR350257-25GEA
78.6
EUR
APOSOR350257-25G
3-(Methoxycarbonyl)benzoesäure
Methyl hydrogen isophthalate
Formel:
C₉H₈O₄ Molekulargewicht: 180,16 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00029972 CAS-Nummer: 1877-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...