2-Isopropoxy-3-pyridinboronsäure 95+%

Lieferant: Apollo Scientific

APOSOR360200-1GEA 68.4 EUR
APOSOR360200-1G
2-Isopropoxy-3-pyridinboronsäure 95+%
2-Isopropoxy-3-pyridinboronsäure
Formel: C₈H₁₂BNO₃
Molekulargewicht: 181,0000 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11855840
CAS-Nummer: 1150114-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR