4-Methyl-2,6-chinolindiol

Achtung

APOSOR3644-1GEA 0 EUR
APOSOR3644-1G APOSOR3644-250MG
4-Methyl-2,6-chinolindiol
4-Methyl-2,6-chinolindiol
Formel: C₁₀H₉NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08437574
CAS-Nummer: 34982-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR