4-(Propoxycarbonyl)phenylboronsäure 97%
APOSOR3966-25GEA
0
EUR
APOSOR3966-25G
APOSOR3966-1G
APOSOR3966-5G
4-(Propoxycarbonyl)phenylboronsäure 97%
4-(Propoxycarbonyl)phenylboronsäure
Formel:
C₁₀H₁₃BO₄ Molekulargewicht: 208,02 g/mol Schmelzpunkt: 108…112 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06659852 CAS-Nummer: 91062-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...