7-Isochinolinboronsäure

Lieferant: Apollo Scientific
Achtung

APOSOR40225-1GEA 287 EUR
APOSOR40225-1G APOSOR40225-250MG
7-Isochinolinboronsäure
7-Isoquinolineboronic acid
Formel: C₉H₈BNO₂
Molekulargewicht: 172,9763 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08234617
CAS-Nummer: 1092790-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR