(2S)-Cyclohexyl({[(2-methyl-2-propanyl)oxy]carbonyl}amino)essigsäure 95%
Lieferant: Apollo Scientific
APOSOR480610-25GEA
171
EUR
APOSOR480610-25G
(2S)-Cyclohexyl({[(2-methyl-2-propanyl)oxy]carbonyl}amino)essigsäure 95%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid
Formel:
C₁₃H₂₃NO₄ Molekulargewicht: 257,3 g/mol Schmelzpunkt: 83 °C Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
109183-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...