9-Phenanthreneboronic acid 98%

Lieferant: Apollo Scientific

APOSOR5963-1GEA 176 EUR
APOSOR5963-1G
9-Phenanthreneboronic acid 98%
9-Phenanthreneboronic acid
Formel: C₁₄H₁₁BO₂
Molekulargewicht: 222,05 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00143524
CAS-Nummer: 68572-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR