2-Amino-4-(4-nitrophenyl)thiazol 97%
Lieferant: Apollo Scientific
APOSOR6122-1GEA
139
EUR
APOSOR6122-1G
2-Amino-4-(4-nitrophenyl)thiazol 97%
2-Amino-4-(4-nitrophenyl)thiazole
Formel:
C₉H₇N₃O₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00170222 CAS-Nummer: 2104-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...