5,6,7,8-Tetrahydrochinoxalin 98%
Lieferant: Apollo Scientific
APOSOR933039-100GEA
248
EUR
APOSOR933039-100G
APOSOR933039-25G
5,6,7,8-Tetrahydrochinoxalin 98%
5,6,7,8-Tetrahydrochinoxalin
Formel:
C₈H₁₀N₂ Molekulargewicht: 134,18 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00038078 CAS-Nummer: 34413-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...