N-Benzylidenanilin 95%
Lieferant: Apollo Scientific
APOSOR936635-25GEA
102
EUR
APOSOR936635-25G
N-Benzylidenanilin 95%
N-Benzylidenanilin
Formel:
C₁₃H₁₁N Molekulargewicht: 181,24 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003027 CAS-Nummer: 538-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...