3-Vinylanisol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methoxy-3-vinylbenzene

OR944915-1G OR944915-25G OR944915-5G
APOSOR944915-1GEA 132 EUR
APOSOR944915-1G APOSOR944915-25G APOSOR944915-5G
3-Vinylanisol 95%
3-Vinylanisol
Formel: C₉H₁₀O
Molekulargewicht: 134,18 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03427262
CAS-Nummer: 626-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR