2-(Trifluormethylthio)benzylamin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
APOSPC2364-1GEA
361
EUR
APOSPC2364-1G
2-(Trifluormethylthio)benzylamin 98%
2-(Trifluormethylthio)benzylamin
Formel:
C₈H₈F₃NS Lagerungstemperatur: Raumtemperatur |
UN-Nummer:
3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...