2-Fluor-5-methyl-4-(trifluormethyl)zimtsäure 98%
Lieferant: Apollo Scientific
APOSPC302245-1GEA
136
EUR
APOSPC302245-1G
2-Fluor-5-methyl-4-(trifluormethyl)zimtsäure 98%
2-Fluor-5-methyl-4-(trifluormethyl)zimtsäure
Formel:
C₁₁H₈F₄O₂ Molekulargewicht: 248,17 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD19687201 CAS-Nummer: 1324063-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...