5-Chlor-2-(trifluormethoxy)benzonitril 98%
Lieferant: Apollo Scientific
APOSPC302613-1GEA
171
EUR
APOSPC302613-1G
5-Chlor-2-(trifluormethoxy)benzonitril 98%
5-Chlor-2-(trifluormethoxy)benzonitril
Formel:
C₈H₃ClF₃NO Molekulargewicht: 221,57 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD11519364 CAS-Nummer: 1092461-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...