2-Chlor-5-fluorisonicotinsäure
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC305019-1GEA
35
EUR
APOSPC305019-1G
2-Chlor-5-fluorisonicotinsäure
2-Chlor-5-fluorisonicotinsäure
Formel:
C₆H₃ClFNO₂ Molekulargewicht: 175,5500 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04972381 CAS-Nummer: 884494-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...