3,4-Difluor-5-(trifluormethyl)benzoesäure 95% min

Lieferant: Apollo Scientific
Achtung

PC3342-250MG
APOSPC3342-250MGEA 147 EUR
APOSPC3342-250MG
3,4-Difluor-5-(trifluormethyl)benzoesäure 95% min
3,4-Difluoro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00236257
CAS-Nummer: 237761-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR