5-(4-Fluorphenyl)-3-pyridincarbonsäure

Lieferant: Apollo Scientific

APOSPC408000-1GEA 372 EUR
APOSPC408000-1G
5-(4-Fluorphenyl)-3-pyridincarbonsäure
5-(4-Fluorophenyl)-3-pyridinecarboxylic acid
Formel: C₁₂H₈FNO₂
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 364064-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR