4-(4'-Fluorbenzyloxy)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC430510-1GEA
149
EUR
APOSPC430510-1G
4-(4'-Fluorbenzyloxy)phenylboronsäure
4-(4'-Fluorbenzyloxy)phenylboronsäure
Formel:
C₁₃H₁₂BFO₃ Molekulargewicht: 246,0420 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07784403 CAS-Nummer: 871125-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...