Ethyl-4-chlor-6-fluorchinolin-3-carboxylat
Lieferant: Apollo Scientific
APOSPC430527-500MGEA
200
EUR
APOSPC430527-500MG
Ethyl-4-chlor-6-fluorchinolin-3-carboxylat
Ethyl-4-chlor-6-fluorchinolin-3-carboxylat
Formel:
C₁₂H₉ClFNO₂ Molekulargewicht: 253,66 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00173350 CAS-Nummer: 77779-49-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...