Kaliumallyltrifluoroborat 95+%
Lieferant: Apollo Scientific
APOSPC48433-25GEA
165
EUR
APOSPC48433-25G
Kaliumallyltrifluoroborat 95+%
Kaliumallyltrifluoroborat
Formel:
C₃H₅BF₃K Molekulargewicht: 147,976 g/mol Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07368222 CAS-Nummer: 233664-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...