2-Fluor-5-(phenylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
APOSPC5105-1GEA
248
EUR
APOSPC5105-1G
2-Fluor-5-(phenylcarbamoyl)phenylboronsäure 98%
2-Fluor-5-(phenylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₁BFNO₃ Molekulargewicht: 259,0400 g/mol Schmelzpunkt: 252…256 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08235095 CAS-Nummer: 874289-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...