3-Brom-4-fluorbenzylamin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC7387-1GEA
16.1
EUR
APOSPC7387-1G
3-Brom-4-fluorbenzylamin Hydrochlorid
3-Brom-4-fluorbenzylamin Hydrochlorid
Formel:
C₇H₈BrClFN Molekulargewicht: 240,50 g/mol Schmelzpunkt: 214…218 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00143094 CAS-Nummer: 202865-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...