2-Brom-4'-fluor-2'-hydroxyacetophenone

Lieferant: Apollo Scientific

Gefahr

APOSPC7605-1GEA 139 EUR
APOSPC7605-1G
2-Brom-4'-fluor-2'-hydroxyacetophenone
2-Brom-4'-fluor-2'-hydroxyacetophenone
Formel: C₈H₆BrFO₂
Lagerungstemperatur: Kühlschrank
UN-Nummer: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR