2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid
Lieferant: Apollo Scientific
APOSPC7979-1GEA
176
EUR
APOSPC7979-1G
2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid
2-Brom-5-fluor-N-(2-hydroxyethyl)benzamid
Formel:
C₉H₉BrFNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09800913 CAS-Nummer: 951884-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...