2-Brom-N-ethyl-5-fluorbenzamid

Lieferant: Apollo Scientific

APOSPC8025-1GEA 176 EUR
APOSPC8025-1G
2-Brom-N-ethyl-5-fluorbenzamid
2-Brom-N-ethyl-5-fluorbenzamid
Formel: C₉H₉BrFNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800905
CAS-Nummer: 951884-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR