Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC8131-1GEA
14.7
EUR
APOSPC8131-1G
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Formel:
C₁₃H₁₀F₃NO₃ Molekulargewicht: 285,2186 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09878378 CAS-Nummer: 23851-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...