N-Cyclopropyl-4-fluor-2-nitroanilin

Lieferant: Apollo Scientific

APOSPC8336-5GEA 379 EUR
APOSPC8336-5G
N-Cyclopropyl-4-fluor-2-nitroanilin
N-Cyclopropyl-4-fluor-2-nitroanilin
Formel: C₉H₉FN₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08448315
CAS-Nummer: 887351-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR