2-(4-Fluorphenyl)oxazol 97%

Lieferant: Apollo Scientific

APOSPC901976-10GEA 887 EUR
APOSPC901976-10G
2-(4-Fluorphenyl)oxazol 97%
2-(4-Fluorophenyl)oxazole
Formel: C₉H₆FNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08234435
CAS-Nummer: 885268-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR