Membrane nitrogen generator, Infinity XE 5030
PEAK3300574EA 21218 EUR
PEAK3300574
Membrane nitrogen generator, Infinity XE 5030
Producing between 30 to 390 L/min of high quality, laboratory grade nitrogen gas, the Infinity XE 503X can comfortably supply gas for multiple laboratory instruments. With variable nitrogen purity available between 95 to 99,5% (depending on flow rate), Infinity XE generators can be configured to suit the needs of several applications, such as LC-MS, ELSD, sample preparation and more.
  • Flow rates from 30 to 390 L/min
  • Variable flow rate, with up to 9,31 bar outlet pressure
  • N₂ purity from 95 to 99,5% (dependent on flow rate)
  • Tried and tested membrane technology for high quality gas
  • Small footprint using minimal laboratory space
  • Effectively silent when in operation
  • O₂ analyser option available, XE 5031 model includes PurityGuard for purity critical applications