Pentair Polypropylene String-Wound Filter Cartridges
Lieferant: Pentair
MFLX29802-42EA 44 EUR
MFLX29802-42 MFLX29802-43 MFLX29802-44 MFLX29802-40 MFLX29802-38 MFLX01508-76 MFLX29802-39 MFLX01508-77 MFLX01508-78 MFLX29802-45 MFLX29802-35 MFLX29802-36 MFLX29802-37
Pentair Polypropylene String-Wound Filter Cartridges
Filter Filterkartuschen
String-wound pattern provides greater surface area for lower pressure drop and higher flow.
  • Contructed of polypropylene (PP)
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165 °F (74 °C).