Two-Way, Three-Way, and Sample Diaphragm Valves
Lieferant: Parker
MFLX98402-08EA 486.41 EUR
MFLX98402-08 MFLX31101-03 MFLX30502-07 MFLX98403-08 MFLX31101-05 MFLX31101-07
Two-Way, Three-Way, and Sample Diaphragm Valves
Ventile Membranventile
Easily control flow with diaphragm valve.
  • Constructed of chemically inert PFA for a metal-free degsign
These diaphragm valves are made of PFA and feature no exposed metal parts. Choose between a two-way, three-way or sample diaphragm valve with various temperature and pressure ranges. Two-way valve 31101-05 includes bracket and screws. Three-way and sample valves include screws for mounting hardware.