Hayward NVA Series Plastic Needle Valves
Lieferant: Hayward
MFLX03245-90EA 171 EUR
MFLX03245-90 MFLX03245-70 MFLX03245-72 MFLX03245-60 MFLX03245-74 MFLX03245-62 MFLX03245-76 MFLX03245-86 MFLX03245-64 MFLX03245-66 MFLX03245-88 MFLX03245-68
Hayward NVA Series Plastic Needle Valves
Ventile Nadelventile
Don’t worry about leaks.
  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
Precision needle valves are available in four materials: PVC, CPVC, PP and PVDF. All feature an integrated stem with PTFE seat; fine pitch stem threads ensure precise adjustment. Valve body also includes flanges for panel mounting. Connections are standard pipe thread.