Reverse phase HPLC columns, Epic C18, Sub 2 µm
Lieferant: ES Industries
Epic®
ESIN522A91-EC18EA 0 EUR
ESIN522A91-EC18 ESIN512A91-EC18
Reverse phase HPLC columns, Epic C18, Sub 2 µm
Chromatographiesäulen
ES industries is pleased to introduce a line of Epic 1,8 µm columns for use with ultra high pressure liquid chromatographs.
These columns have been engineered to specifically for chromatography at pressures up to 19000 psi. ES Industries expertise in the silica surface modification and bond stationary technology has enabled us to develop a number of stationary phases based on the 1,8 µm particle technology. With this breakthrough the chromatographer is not limited to ODS type phases. The Epic 1,8 µm columns will provide the chromatographer with the highest resolution over a wide variety flow rate conditions and mobile compositions.