N-Acryloylamido-ethoxyethanol ~50% in Wasser ≥98,0%, Fluka™

Lieferant: Honeywell Chemicals

HONC12059-50MLEA 4535.49 EUR
HONC12059-50ML
N-Acryloylamido-ethoxyethanol ~50% in Wasser ≥98,0%, Fluka™
N-Acryloylamido-ethoxyethanol
Formel: C₇H₁₃NO₃
Molekulargewicht: 159,18 g/mol
Dichte: 1,075 g/cm3 (20 °C)
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 89911-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR