Amino-PEG5-acid 96%

BLDPBD00845171-50MEA 150 EUR
BLDPBD00845171-50M
Amino-PEG5-acid 96%
Amino-PEG5-acid

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formel: C₁₃H₂₇NO₇
Molekulargewicht: 309,356 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD25424126
CAS-Nummer: 1191078-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR