(S)-N-Cbz-2-amino-1-phenylethylamine 95%

BLDPBD00849767-1GEA 495 EUR
BLDPBD00849767-1G BLDPBD00849767-250 BLDPBD00849767-100
(S)-N-Cbz-2-amino-1-phenylethylamine 95%
(S)-N-Cbz-2-amino-1-phenylethylamine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₆H₁₈N₂O₂
Molekulargewicht: 270,326 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09260584
CAS-Nummer: 130406-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR